Gene Transcripts

Gene Symbol
Rc3h2
Entrez Gene ID
319817
Gene Name
ring finger and CCCH-type zinc finger domains 2
Gene Aliases
2900024N03Rik, 9430019J22Rik, D930043C02Rik, Mnab, Rnf164
Chromosome Location
Chr. 2: 37368861 - 37423380 on Build GRCm38
UniGene ID
Mm.288785
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100591.1 NP_001094061.1 22 8475
NM_001290642.1 NP_001277571.1 22 8456
GenBank mRNA AK013592.1 - 1 1080
BC006655.1 - 1 1805
BC031826.1 - 1 1829

View Full Product Details