Gene Transcripts

Gene Symbol
Ism1
Entrez Gene ID
319909
Gene Name
isthmin 1 homolog (zebrafish)
Gene Aliases
5430433G21Rik, Ism
Chromosome Location
Chr. 2: 139675007 - 139758581 on Build GRCm38
UniGene ID
Mm.41113
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276489.1 NP_001263418.1 6 2524
GenBank mRNA AK077398.1 - 6 2526

View Full Product Details