Gene Transcripts

Gene Symbol
Cacnb1
Entrez Gene ID
50688
Gene Name
calcium channel, voltage-dependent, beta 1 subunit
Gene Aliases
Chromosome Location
Chr. 10: 85954138 - 85974777 on Build Rnor_6.0
UniGene ID
Rn.9417
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017346.1 NP_059042.1 1, 2 155
XM_006247409.2 XP_006247471.1 1, 2 288
GenBank mRNA X61394.1 - 1, 2 155

View Full Product Details