Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001033453.3 | NP_001028625.1 | 2 | 3733 | |
NM_001098230.1 | NP_001091700.1 | 3 | 3768 | ||
NM_001098231.1 | NP_001091701.1 | 2 | 3684 | ||
NM_001290387.1 | NP_001277316.1 | 2 | 3682 | ||
NM_001290391.1 | NP_001277320.1 | 3 | 3717 | ||
GenBank mRNA | AK133542.1 | - | 2 | 3676 | |
BB077113.2 | - | 1 | 205 |