Gene Transcripts

Gene Symbol
Foxr1
Entrez Gene ID
382074
Gene Name
forkhead box R1
Gene Aliases
Foxn5, Gm1115
Chromosome Location
Chr. 9: 44434234 - 44440868 on Build GRCm38
UniGene ID
Mm.335103
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033469.2 NP_001028641.1 5 1121
GenBank mRNA AK139562.1 - 5 1121

View Full Product Details