Gene Transcripts

Gene Symbol
Brwd3
Entrez Gene ID
382236
Gene Name
bromodomain and WD repeat domain containing 3
Gene Aliases
Brodl, D030064D06Rik, Gm596
Chromosome Location
Chr. X: 108737225 - 108834360 on Build GRCm38
UniGene ID
Mm.100112
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081477.1 NP_001074946.1 1, 2 262
GenBank mRNA AK042444.1 - 254
AK050539.1 - 1, 2 248
AK163354.1 - 1, 2 254

View Full Product Details