Gene Transcripts

Gene Symbol
Cdkl5
Entrez Gene ID
382253
Gene Name
cyclin-dependent kinase-like 5
Gene Aliases
BC038161, Stk9
Chromosome Location
Chr. X: 160784308 - 160994681 on Build GRCm38
UniGene ID
Mm.443717
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024624.2 NP_001019795.1 5, 6 534
GenBank mRNA AK052380.1 - 5, 6 531
AK139220.1 - 5, 6 506

View Full Product Details