Gene Transcripts

Gene Symbol
Tmed8
Entrez Gene ID
382620
Gene Name
transmembrane emp24 domain containing 8
Gene Aliases
6430595O10Rik, AI447224, Gm1184, Mem1
Chromosome Location
Chr. 12: 87166242 - 87200229 on Build GRCm38
UniGene ID
Mm.374912
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033475.3 NP_001028647.1 6 6676
GenBank mRNA AK018286.1 - 1 682
AK147378.1 - 6 6677

View Full Product Details