Gene Transcripts

Gene Symbol
Frmd7
Entrez Gene ID
385354
Gene Name
FERM domain containing 7
Gene Aliases
EG665849, Gm1533
Chromosome Location
Chr. X: 50892645 - 50943619 on Build GRCm38
UniGene ID
Mm.393050
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190332.1 NP_001177261.1 12 3213
GenBank mRNA AK032347.1 - 1 1544

View Full Product Details