Gene Transcripts

Gene Symbol
Mn1
Entrez Gene ID
433938
Gene Name
meningioma 1
Gene Aliases
AA003644, AA009236
Chromosome Location
Chr. 5: 111418166 - 111457025 on Build GRCm38
UniGene ID
Mm.332576
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081235.1 NP_001074704.1 2 5776
GenBank mRNA AK142207.1 - 4888

View Full Product Details