Gene Transcripts

Gene Symbol
Ldlrad2
Entrez Gene ID
435811
Gene Name
low density lipoprotein receptor A domain containing 2
Gene Aliases
Chromosome Location
Chr. 4: 137570876 - 137582385 on Build GRCm38
UniGene ID
Mm.359646
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033979.1 NP_001029151.1 4 1156
GenBank mRNA AK132881.1 - 4 1156

View Full Product Details