Gene Transcripts

Gene Symbol
Gpr62
Entrez Gene ID
436090
Gene Name
G protein-coupled receptor 62
Gene Aliases
4933402E03Rik
Chromosome Location
Chr. 9: 106463960 - 106466612 on Build GRCm38
UniGene ID
Mm.482905
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159652.1 NP_001153124.1 1 1932
GenBank mRNA AK005968.1 - 1 923
AK016613.1 - 1 910

View Full Product Details