Gene Transcripts

Gene Symbol
Ubtfl1
Entrez Gene ID
546118
Gene Name
upstream binding transcription factor, RNA polymerase I-like 1
Gene Aliases
B020006M18Rik, Hmgpi
Chromosome Location
Chr. 9: 18404418 - 18411502 on Build GRCm38
UniGene ID
Mm.380138
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033793.3 NP_001028965.1 1 91
GenBank mRNA AK139937.1 - 1 92
AK163257.1 - 1 90

View Full Product Details