Gene Transcripts
- Gene Symbol
- Gata6
- Entrez Gene ID
- 29300
- Gene Name
- GATA binding protein 6
- Gene Aliases
- Chromosome Location
- Chr. 18: 2416871 - 2446338 on Build Rnor_6.0
- UniGene ID
- Rn.8701
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019185.1 |
NP_062058.1
|
4 |
1143 |
|
GenBank mRNA |
L22760.1 |
- |
4 |
1143 |
|
View Full Product Details