Gene Transcripts

Gene Symbol
Vmn1r31
Entrez Gene ID
626828
Gene Name
vomeronasal 1 receptor 31
Gene Aliases
EG626828, Gm6709
Chromosome Location
Chr. 6: 58471967 - 58472878 on Build GRCm38
UniGene ID
Mm.480268
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166729.1 NP_001160201.1 1 193

View Full Product Details