Gene Transcripts

Gene Symbol
Fbxo39
Entrez Gene ID
628100
Gene Name
F-box protein 39
Gene Aliases
1700010H23Rik
Chromosome Location
Chr. 11: 72314370 - 72319419 on Build GRCm38
UniGene ID
Mm.390365
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099688.2 NP_001093158.1 2 313
GenBank mRNA BU937284.1 - 2 316

View Full Product Details