Gene Transcripts

Gene Symbol
Samd1
Entrez Gene ID
666704
Gene Name
sterile alpha motif domain containing 1
Gene Aliases
AA589507
Chromosome Location
Chr. 8: 83997672 - 84000386 on Build GRCm38
UniGene ID
Mm.298091
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081415.1 NP_001074884.1 2 1171


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA W83690.1 - 90 Chr. - - [-]

View Full Product Details