Gene Transcripts

Gene Symbol
Sox10
Entrez Gene ID
29361
Gene Name
SRY (sex determining region Y)-box 10
Gene Aliases
Chromosome Location
Chr. 7: 120393254 - 120403523 on Build Rnor_6.0
UniGene ID
Rn.10883
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019193.2 NP_062066.1 5 1348
GenBank mRNA AJ001029.1 - 5 1349
BC062067.1 - 4 1060 5599259

View Full Product Details