Gene Transcripts
- Gene Symbol
- Vmn2r88
- Entrez Gene ID
- 669149
- Gene Name
- vomeronasal 2, receptor 88
- Gene Aliases
- V2r13, V2r3
- Chromosome Location
- Chr. 14: 51411001 - 51419527 on Build GRCm38
- UniGene ID
- Mm.377117
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_011686.1 |
NP_035816.1
|
2 |
497 |
|
View Full Product Details