Gene Transcripts

Gene Symbol
H60c
Entrez Gene ID
670558
Gene Name
histocompatibility 60c
Gene Aliases
4632413I24Rik, Gm9491
Chromosome Location
Chr. 10: 3256202 - 3268092 on Build GRCm38
UniGene ID
Mm.398582
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204916.2 NP_001191845.1 6 1008
GenBank mRNA AB284506.1 - 6 987
AK014573.1 - 3 575

View Full Product Details