Gene Transcripts

Gene Symbol
Dtwd2
Entrez Gene ID
68857
Gene Name
DTW domain containing 2
Gene Aliases
1190002H09Rik, 8030470C17Rik, AI414673, BB115449
Chromosome Location
Chr. 18: 49696145 - 49755667 on Build GRCm38
UniGene ID
Mm.96010
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170960.1 NP_001164431.1 4 1631
NM_026854.3 NP_081130.1 6 1919
GenBank mRNA AK033232.1 - 1 166
AK144689.1 - 6 1891

View Full Product Details