Gene Transcripts

Gene Symbol
Izumo3
Entrez Gene ID
69314
Gene Name
IZUMO family member 3
Gene Aliases
1700011H22Rik
Chromosome Location
Chr. 4: 92144318 - 92147221 on Build GRCm38
UniGene ID
Mm.55891
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027034.1 NP_081310.1 7 1170
GenBank mRNA AK005867.1 - 7 718
BC099579.1 - 7 1169

View Full Product Details