Gene Transcripts

Gene Symbol
Tmem89
Entrez Gene ID
69384
Gene Name
transmembrane protein 89
Gene Aliases
1700024B07Rik
Chromosome Location
Chr. 9: 108914619 - 108915563 on Build GRCm38
UniGene ID
Mm.158271
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027066.1 NP_081342.1 1 51
GenBank mRNA AK006292.1 - 1 53
BC111913.1 - 1 51 40053443
BY706371.1 - 1 53

View Full Product Details