Gene Transcripts

Gene Symbol
Krtap3-1
Entrez Gene ID
69473
Gene Name
keratin associated protein 3-1
Gene Aliases
2310003K17Rik, KAP3.1, KRTAP3.1
Chromosome Location
Chr. 11: 99566027 - 99566630 on Build GRCm38
UniGene ID
Mm.158281
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023511.1 NP_076000.1 1 291
GenBank mRNA AK009119.1 - 1 41
D86424.1 - 1 239

View Full Product Details