Gene Transcripts

Gene Symbol
Enho
Entrez Gene ID
69638
Gene Name
energy homeostasis associated
Gene Aliases
1110065P19Rik, 2310040A07Rik
Chromosome Location
Chr. 4: 41638144 - 41640302 on Build GRCm38
UniGene ID
Mm.372459
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027147.1 NP_081423.1 2 1152
GenBank mRNA AK009710.1 - 2 1152
AK050496.1 - 2 951
AK182884.1 - 1 299
AK205625.1 - 1 299
AK206064.1 - 1 298
AK206108.1 - 1 299
AK207085.1 - 1 284
AK208424.1 - 1 299
AK214699.1 - 1 299
AK216096.1 - 1 299
AK219775.1 - 1 293
BC021944.1 - 2 810

View Full Product Details