Gene Transcripts

Gene Symbol
Naa30
Entrez Gene ID
70646
Gene Name
N(alpha)-acetyltransferase 30, NatC catalytic subunit
Gene Aliases
4930487N19Rik, 5730533P17Rik, AI447922, AW322455, Nat12
Chromosome Location
Chr. 14: 49172133 - 49191031 on Build GRCm38
UniGene ID
Mm.275688
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081430.1 NP_001074899.1 5 2432
GenBank mRNA AK017805.1 - 1 243
AK044235.1 - 4 2052
BC054060.1 - 3 1659

View Full Product Details