Gene Transcripts

Gene Symbol
Fam47c
Entrez Gene ID
70864
Gene Name
family with sequence similarity 47, member C
Gene Aliases
4921509A18Rik, Fam47a, Fam47a-ps
Chromosome Location
Chr. X: 78737763 - 78739410 on Build GRCm38
UniGene ID
Mm.66681
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164739.1 NP_001158211.1 1 1355
GenBank mRNA AK014848.1 - 1 1351
BC115971.1 - 1 1326 40106467

View Full Product Details