Gene Transcripts

Gene Symbol
Armc3
Entrez Gene ID
70882
Gene Name
armadillo repeat containing 3
Gene Aliases
4921513G22Rik
Chromosome Location
Chr. 2: 19198865 - 19310243 on Build GRCm38
UniGene ID
Mm.276696
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081083.2 NP_001074552.1 16, 17 2269
GenBank mRNA AK014880.1 - 16, 17 2085

View Full Product Details