Gene Transcripts

Gene Symbol
Naa40
Entrez Gene ID
70999
Gene Name
N(alpha)-acetyltransferase 40, NatD catalytic subunit, homolog (S. cerevisiae)
Gene Aliases
4931433E08Rik, AU023197, Nat11
Chromosome Location
Chr. 19: 7225668 - 7241222 on Build GRCm38
UniGene ID
Mm.329605
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027643.1 NP_081919.1 8 1454
GenBank mRNA AK016504.1 - 8 1415
AK033510.1 - 8 1276
AK033896.1 - 8 1285
AK035418.1 - 7 2172
AK162625.1 - 7 1548
AK170548.1 - 8 1280
AK187331.1 - 1 72
BC020020.1 - 8 1305 3156924
BC049808.1 - 2 692
BC058212.1 - 3 1113

View Full Product Details