Gene Transcripts

Gene Symbol
Krtap16-3
Entrez Gene ID
71369
Gene Name
keratin associated protein 16-3
Gene Aliases
5430404J20Rik, Krtap16-10
Chromosome Location
Chr. 16: 88962304 - 88962873 on Build GRCm38
UniGene ID
Mm.251757
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183296.1 NP_899119.1 1 527
GenBank mRNA AF345293.1 - 1 467
AK017265.1 - 1 527

View Full Product Details