Gene Transcripts

Gene Symbol
Shcbp1l
Entrez Gene ID
71836
Gene Name
Shc SH2-domain binding protein 1-like
Gene Aliases
1700012A16Rik
Chromosome Location
Chr. 1: 153425098 - 153452574 on Build GRCm38
UniGene ID
Mm.49460
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033162.2 NP_001028334.1 7 1232
GenBank mRNA AK005886.1 - 7 1229
AK161274.1 - 7 1233
BC113157.1 - 7 1279 40058375

View Full Product Details