Gene Transcripts

Gene Symbol
Efhc1
Entrez Gene ID
71877
Gene Name
EF-hand domain (C-terminal) containing 1
Gene Aliases
1700029F22Rik, mRib72-1, myoclonin1
Chromosome Location
Chr. 1: 20951551 - 20990841 on Build GRCm38
UniGene ID
Mm.29178
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027974.1 NP_082250.1 6 1170
GenBank mRNA AK006489.1 - 6 1109
EU520262.1 - 6 1062

View Full Product Details