Gene Transcripts
- Gene Symbol
- Efhc1
- Entrez Gene ID
- 71877
- Gene Name
- EF-hand domain (C-terminal) containing 1
- Gene Aliases
- 1700029F22Rik, mRib72-1, myoclonin1
- Chromosome Location
- Chr. 1: 20951551 - 20990841 on Build GRCm38
- UniGene ID
- Mm.29178
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_027974.1 |
NP_082250.1
|
11 |
2128 |
|
GenBank mRNA |
AK006489.1 |
- |
11 |
2067 |
|
View Full Product Details