Gene Transcripts

Gene Symbol
Itpripl1
Entrez Gene ID
73338
Gene Name
inositol 1,4,5-triphosphate receptor interacting protein-like 1
Gene Aliases
1700041B20Rik
Chromosome Location
Chr. 2: 127138769 - 127143488 on Build GRCm38
UniGene ID
Mm.57559
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163527.1 NP_001156999.1 2 3605
NM_001163528.1 NP_001157000.1 2 3429
GenBank mRNA AK028913.1 - 3 3340

View Full Product Details