Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019222.1 | NP_062095.1 | 8 | 1014 | |
XM_006230773.2 | XP_006230835.1 | 8 | 1133 | ||
XM_006230774.2 | XP_006230836.1 | 8 | 1098 | ||
GenBank mRNA | AJ006064.1 | - | 8 | 1014 | |
BC061558.1 | - | 7 | 1612 | 6919414 | |
FQ213235.1 | - | 4 | 387 |