Gene Transcripts

Gene Symbol
Wbp2nl
Entrez Gene ID
74716
Gene Name
WBP2 N-terminal like
Gene Aliases
4930521I23Rik, PAWP
Chromosome Location
Chr. 15: 82298984 - 82314558 on Build GRCm38
UniGene ID
Mm.159315
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029066.1 NP_083342.1 3 233
GenBank mRNA AK015863.1 - 3 264
BC119520.2 - 3 233 40055925

View Full Product Details