Gene Transcripts

Gene Symbol
Etnk1
Entrez Gene ID
75320
Gene Name
ethanolamine kinase 1
Gene Aliases
1110061E11Rik, 4930555L11Rik, AI195356, AI841245, C80956, D6Ertd3e, EKI1
Chromosome Location
Chr. 6: 143166866 - 143208546 on Build GRCm38
UniGene ID
Mm.272548
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029250.2 NP_083526.2 3 750
GenBank mRNA AK016135.1 - 3 732
AK031560.1 - 727
AK033268.1 - 3 727
AK044502.1 - 3 751
AK049827.1 - 3 464
AK051039.1 - 760
AK164928.1 - 3 727

View Full Product Details