Gene Transcripts

Gene Symbol
Dnajc21
Entrez Gene ID
78244
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 21
Gene Aliases
4930461P20Rik, 9930116P15Rik
Chromosome Location
Chr. 15: 10446763 - 10470516 on Build GRCm38
UniGene ID
Mm.297862
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030046.2 NP_084322.2 5 786
GenBank mRNA AK141303.1 - 2 246

View Full Product Details