Gene Transcripts

Gene Symbol
Extl3
Entrez Gene ID
56819
Gene Name
exostosin-like glycosyltransferase 3
Gene Aliases
Regr
Chromosome Location
Chr. 15: 48420419 - 48465318 on Build Rnor_6.0
UniGene ID
Rn.48671
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020097.1 NP_064482.1 1 315
XM_006252206.2 XP_006252268.1 4 1621
GenBank mRNA AB033367.1 - 1 315

View Full Product Details