Gene Transcripts

Gene Symbol
Itih4
Entrez Gene ID
54404
Gene Name
inter-alpha-trypsin inhibitor heavy chain family, member 4
Gene Aliases
Chromosome Location
Chr. 16: 6970367 - 6985538 on Build Rnor_6.0
UniGene ID
Rn.11308
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019369.2 NP_062242.2 22 2690
GenBank mRNA BC089806.1 - 22 2690
FQ210475.1 - 22 2646
Y11283.1 - 22 2687

View Full Product Details