Gene Transcripts

Gene Symbol
Gorasp1
Entrez Gene ID
56082
Gene Name
golgi reassembly stacking protein 1
Gene Aliases
GRASP65
Chromosome Location
Chr. 8: 128610290 - 128622337 on Build Rnor_6.0
UniGene ID
Rn.144458
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019385.2 NP_062258.2 3 184
GenBank mRNA AF015264.2 - 3 161
BC081809.1 - 3 178
BC161850.1 - 3 184 8369057

View Full Product Details