Gene Transcripts

Gene Symbol
Ltbp2
Entrez Gene ID
59106
Gene Name
latent transforming growth factor beta binding protein 2
Gene Aliases
Chromosome Location
Chr. 6: 108500114 - 108596638 on Build Rnor_6.0
UniGene ID
Rn.40921
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021586.2 NP_067597.2 1 324
XM_006240350.2 XP_006240412.1 1 516
GenBank mRNA DV213994.1 - 1 426
Y12760.1 - 1 323

View Full Product Details