Gene Transcripts

Gene Symbol
Aipl1
Entrez Gene ID
59110
Gene Name
aryl hydrocarbon receptor-interacting protein-like 1
Gene Aliases
Chromosome Location
Chr. 10: 58579129 - 58631194 on Build Rnor_6.0
UniGene ID
Rn.8243
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021590.1 NP_067601.1 4 562
XM_006246798.2 XP_006246860.1 4 594
GenBank mRNA AF180340.1 - 4 562

View Full Product Details