Gene Transcripts

Gene Symbol
Fgf7
Entrez Gene ID
29348
Gene Name
fibroblast growth factor 7
Gene Aliases
Kgf
Chromosome Location
Chr. 3: 118315856 - 118368464 on Build Rnor_6.0
UniGene ID
Rn.98842
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022182.1 NP_071518.1 3 642
XM_006234903.2 XP_006234965.1 4 1962
GenBank mRNA AF295300.1 - 3 610
X56551.1 - 3 642

View Full Product Details