Gene Transcripts

Gene Symbol
Khdrbs3
Entrez Gene ID
64015
Gene Name
KH domain containing, RNA binding, signal transduction associated 3
Gene Aliases
Etle, Slm2
Chromosome Location
Chr. 7: 110031819 - 110179475 on Build Rnor_6.0
UniGene ID
Rn.229046
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022249.1 NP_071585.1 2 236
GenBank mRNA AF152547.1 - 2 236

View Full Product Details