Gene Transcripts

Gene Symbol
Pi4ka
Entrez Gene ID
64161
Gene Name
phosphatidylinositol 4-kinase, catalytic, alpha
Gene Aliases
Pik4ca
Chromosome Location
Chr. 11: 87858309 - 87974394 on Build Rnor_6.0
UniGene ID
Rn.11015
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022301.1 NP_071637.1 6 691
XM_008768902.1 XP_008767124.1 6 835
GenBank mRNA D83538.1 - 6 969

View Full Product Details