Gene Transcripts

Gene Symbol
Srpx
Entrez Gene ID
64316
Gene Name
sushi-repeat-containing protein, X-linked
Gene Aliases
Drs
Chromosome Location
Chr. X: 14150031 - 14220751 on Build Rnor_6.0
UniGene ID
Rn.1489
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022524.1 NP_071969.1 9 1239
XM_006256683.2 XP_006256745.1 8 1267
XM_008773077.1 XP_008771299.1 8 2393
GenBank mRNA BC087639.1 - 8 1238
D78359.1 - 9 1239

View Full Product Details