Gene Transcripts

Gene Symbol
Mvp
Entrez Gene ID
64681
Gene Name
major vault protein
Gene Aliases
Chromosome Location
Chr. 1: 198420821 - 198448418 on Build Rnor_6.0
UniGene ID
Rn.10028
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022715.2 NP_073206.2 2 156
GenBank mRNA BC071174.1 - 2 163
U09870.2 - 2 156

View Full Product Details