Gene Transcripts

Gene Symbol
Rps14
Entrez Gene ID
29284
Gene Name
ribosomal protein S14
Gene Aliases
Chromosome Location
Chr. 18: 56042532 - 56047316 on Build Rnor_6.0
UniGene ID
Rn.154712
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022672.1 NP_073163.1 3 227
GenBank mRNA BC058472.1 - 2 234
FQ210177.1 - 3 229
FQ210178.1 - 3 229
FQ210421.1 - 3 229
FQ210469.1 - 3 226
FQ211405.1 - 3 227
FQ211449.1 - 3 227
FQ211623.1 - 3 229
FQ212082.1 - 3 231
FQ212093.1 - 3 227
FQ214636.1 - 3 229
FQ217229.1 - 3 227
FQ217605.1 - 3 227
FQ218082.1 - 3 224
FQ219972.1 - 3 227
FQ220057.1 - 3 227
FQ220332.1 - 3 229
FQ220676.1 - 3 231
FQ220975.1 - 3 224
FQ221057.1 - 3 227
FQ221058.1 - 3 227
FQ221066.1 - 3 227
FQ221125.1 - 3 229
FQ221205.1 - 3 227
FQ221325.1 - 3 227
FQ221375.1 - 3 227
FQ221450.1 - 3 227
FQ221468.1 - 3 227
FQ221581.1 - 3 226
FQ221634.1 - 3 224
FQ221754.1 - 3 223
FQ221769.1 - 3 229
FQ221835.1 - 3 229
FQ221893.1 - 3 229
FQ221920.1 - 3 229
FQ222066.1 - 3 224
FQ222119.1 - 3 227
FQ222124.1 - 3 227
FQ222163.1 - 3 228
FQ222324.1 - 3 231
FQ222370.1 - 3 227
FQ222480.1 - 3 227
FQ222552.1 - 3 226
FQ222574.1 - 3 227
FQ222575.1 - 2 227
FQ222601.1 - 3 242
FQ222667.1 - 3 227
FQ222685.1 - 3 227
FQ222697.1 - 3 229
FQ222714.1 - 3 227
FQ222747.1 - 3 231
FQ222803.1 - 3 227
FQ222840.1 - 3 230
FQ222893.1 - 3 229
FQ222979.1 - 3 229
FQ222986.1 - 3 224
FQ223007.1 - 3 227
FQ223012.1 - 3 229
FQ223069.1 - 3 229
FQ223131.1 - 3 227
FQ223204.1 - 3 229
FQ223283.1 - 3 227
FQ223454.1 - 3 228
FQ223468.1 - 3 227
FQ223480.1 - 3 227
FQ223519.1 - 3 226
FQ223527.1 - 3 227
FQ223570.1 - 3 227
FQ224051.1 - 3 224
FQ224105.1 - 3 229
FQ224302.1 - 3 228
FQ224808.1 - 3 231
FQ225464.1 - 3 229
FQ228360.1 - 3 212
FQ228480.1 - 3 229
FQ228486.1 - 3 227
FQ228559.1 - 3 231
FQ228608.1 - 3 227
FQ228692.1 - 3 226
FQ229006.1 - 3 233
FQ229014.1 - 3 229
FQ229055.1 - 3 229
FQ229061.1 - 3 229
FQ229229.1 - 3 229
FQ229387.1 - 3 227
FQ231306.1 - 370
X15040.1 - 3 227

Gene Symbol
LOC100911847
Entrez Gene ID
100911847
Gene Name
40S ribosomal protein S14-like
Gene Aliases
Chromosome Location
Chr. 5: 144083043 - 144083596 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750030.3 XP_003750078.3 1 231

View Full Product Details