Gene Transcripts

Gene Symbol
Axin1
Entrez Gene ID
79257
Gene Name
axin 1
Gene Aliases
Axin
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.31781
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024405.1 NP_077381.1 2410
GenBank mRNA AF017756.1 - 2410

View Full Product Details